Learn More
Poltex 6 Pocket PETG Serological Pipette Holder
Click to view available options
Mounting:
Countertop
Magnet Mount
Tape Mount
Includes:
Holder Only
Pipette Controller Bracket
Description
- Serological Pipette (pipet) Holder.
- Choose from 4, 5, or 6 pocket capacity.
- Choose from powerful, interchangeable magnet mount, 3M VHB (very high bond) tape mount, or countertop placement.
- Add a pipette filler bracket to any version (bracket mounts to either side).
- Magnets and tape mount to either side.
- Break-resistant PETG construction.
- PETG has good alcohol/chemical resistance.
- Shelves are removable for easy cleaning.
Specifications
Specifications
Product Type | Serological Pipet Holder |
Includes | Holder Only |
Material | PETG |
Positions | 6 Pockets |
Mounting | Tape Mount |