Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Advanced Instruments Petri Dish Holders

Advanced Instruments Petri Dish Holders available in a variety of sizes, and types for all laboratory needs

$54.49 - $147.73

Specifications

Item Description Stainless Steel
Storage Requirements 50 to 91°F
View More Specs

Products 6
Catalog Number Mfr. No. Size Format Price Quantity & Availability  
Catalog Number Mfr. No. Size Format Price Quantity & Availability  
22-047-622
View Documents
Advanced Instruments
PH1080
6 cm Petri Dishes 3 Stacks
Each for $147.73
Add to cart
 
22-047-620
View Documents
Advanced Instruments
PH1060
9-10 cm Petri Dishes Short Stack
Each for $54.49
Add to cart
 
22-047-618
View Documents
Advanced Instruments
PH1040
9-10 cm Petri Dishes Single Stack
Each for $55.57
Add to cart
 
22-047-619
View Documents
Advanced Instruments
PH1050
10 cm Petri Dishes 3 Stacks
Each for $142.24
Add to cart
 
22-047-621
View Documents
Advanced Instruments
PH1070
14.5 cm Petri Dishes Single Stack
Each for $65.01
Add to cart
 
22-047-623
View Documents
Advanced Instruments
PH1090
13 x 9 cm Microtiter Plates Single Stack
Each for $104.10
Add to cart
 
Description

Description

  • Built with long lasting, durable stainless steel
  • Choose from a variety of sizes that can stack between 6 and 12 Petri dishes/Microtiter Plates
  • Helps to organize and maintain the safety of samples
Specifications

Specifications

Stainless Steel
50 to 91°F
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit