Learn More
Bellco Glass, Inc. Provides automatic switching from the main tank to auxiliary tank.. Rear panel hose barbs are for gas connection. Alarm will sound when tank pressure drops below 5psi. Manual switching is done via the button labeled tank switch. Mounts on top of any stand
SDP
Supplier:  Bellco Glass, Inc. 672700115
Encompass_Preferred
Provides automatic switching from the main tank to auxiliary tank.. Rear panel hose barbs are for gas connection. Alarm will sound when tank pressure drops below 5psi. Manual switching is done via the button labeled tank switch. Mounts on top of any stand
Catalog No. 50-194-5836
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.