Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher Science Education™ Petroleum Ether
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
ACS Grade
Description
This product is for educational use - no C of A is available.
Specifications
Grade | ACS Reagent |
Quantity | 100 mL |
Packaging | ACS Grade |
Chemical Name or Material | Petroleum Ether |
Safety and Handling
ShelfLife | 5 years |
DOTInformation | DOT Class 3, II, : Flammable Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction