Petroleum ether 35/60, Environmental Grade, Alfa Aesar™

Suitable For Environmental and Pesticide Analysis

Manufacturer: Alfa Aesar 40982M1

Quantity 4 x 1L

 View more versions of this product

Catalog No. AA40982M1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 8032-32-4
    Physical Form Liquid
    Assay Percent Range >98%
    MDL Number MFCD00081849
    Chemical Name or Material Petroleum ether 35/60, Environmental Grade
    Grade Environmental Grade

    Specifications

    Physical Form Liquid
    Color Clear colorless
    Quantity 4 x 1L
    DOT Information DOT Class: 3, Packaging Group: I
    EINECS Number 232-453-7
    Ignition Point 250°C (482°F)
    Odor Petroleum-like
    Vapor Pressure 350 hPa (263mm Hg) at 20°C
    Appearance Colorless liquid
    Assay Percent Range >98%
    MDL Number MFCD00081849
    CAS 8032-32-4
    Health Hazard 1 H224-H340-H350-H304: Extremely flammable liquid and vapour. May cause genetic defects. May cause cancer. May be fatal if swallowed and enters airways.
    Chemical Name or Material Petroleum ether 35/60, Environmental Grade
    Grade Environmental Grade

    H224-H340-H350-H304: Extremely flammable liquid and vapour. May cause genetic defects. May cause cancer. May be fatal if swallowed and enters airways.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit