Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Petroleum ether, for analysis, boiling range 80-110°C, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
10L
Packaging:
Glass Bottle
Plastic Jerrycan
64742-49-0
C6H14
86.178
AFABGHUZZDYHJO-UHFFFAOYSA-N
isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane
7892
2-methylpentane
CCCC(C)C
64742-49-0
C6H14
86.178
AFABGHUZZDYHJO-UHFFFAOYSA-N
isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane
7892
2-methylpentane
CCCC(C)C
For analysis
Aromatic Substance 0.01% max.
Benzene 20ppm max.
Cadmium (Cd) 0.1ppm max.
Physical Form Liquid
Grade Analysis
Cobalt (Co) 0.1ppm max.
Copper (Cu) 0.1ppm max.
Density 0.7250g/mL
Infrared Spectrum Authentic
Iron (Fe) 0.2ppm max.
Lead (Pb) 0.1ppm max.
n_Hexane 5% max.
Nickel (Ni) 0.1ppm max.
Packaging Glass Bottle
Refractive Index 1.3980 to 1.4060 (20°C, 589nm)
Zinc (Zn) 0.1ppm max.
Boiling Point 78°C to 113°C
Color Undesignated
Quantity 1L
Chemical Name or Material Petroleum ether
Show More Show Less