Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Petroleum Ether, B.R. 35 DEG -60°C, PestiSolv(R), Spectrum™

Click to view available options
Quantity:
4 L
Packaging:
Amber Glass Bottle
8032-32-4
8032-32-4
CAS Max % 1
Boiling Range 35° to 60°C
Grade High Purity Solvent
Packaging Amber Glass Bottle
Residue after Evaporation 3 ppm
Water 0.0002
Quantity 4 L

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.