Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Petroleum ether, pure, boiling range ca. 180-280°C, Acros Organics

Click to view available options
Quantity:
1L
2.5L
5L
25L
Packaging:
Glass Bottle
Metal drum
Plastic drum
8008-20-6
MFCD00135561
8008-20-6
MFCD00135561
Quantity 1L
Specific Gravity 0.79
Density 0.7900g/mL
Infrared Spectrum Authentic
Boiling Point 200.0°C to 250.0°C
Flash Point 82°C
Color Colorless
Packaging Glass Bottle
Refractive Index 1.4300 to 1.4500
Merck Index 15,5342
Solubility Information Solubility in water: insoluble
Physical Form Liquid
Grade Pure
Chemical Name or Material Petroleum ether
Show More Show Less

GHS H Statement
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.

GHS Signal Word: Danger

EINECSNumber : 232-366-4