Learn More
Petzl™ America CONNEXION FAST Anchor Strap
Manufacturer: Petzl™ C42 V
Specifications
0.47kg | |
31.49 to 51.18 in. | |
CE EN 795 B, CE EN 354 | |
Adjustable anchor strap |
1.04 lb. | |
80 to 130 cm | |
5000 lbf. |