Petzl™ America CORDEX PLUS Mid-weight Belay/Rappel Gloves

Manufacturer: Petzl AmericaSupplier Diversity Partner K53 XLN

 View more versions of this product

Catalog No. 19-130-5641

  Check Availability
Add to Cart

    Specifications

    Certifications CE EN 420, CE EN 388, 4143
    Glove Cuff Style Hook and loop strap
    For Use With (Application) repelling or belaying
    Material Leather, Neoprene
    Size X-Large
    Color Black

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit