Learn More
Petzl™ Shunt Rope Clamp Non-distribution product as customer accommodation.
Manufacturer:  Petzl™ B03B
Catalog No. 19-167-243
Add to cart
Description
Description
Specifications
Specifications
Single ropes 10-11 mm or double ropes 8-11 mm
188 g
Back-up Rope Clamp
Aluminum