Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PEX11A Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$149.00 - $329.00

Specifications

Antigen PEX11A
Immunogen Synthetic peptides corresponding to PEX11A(peroxisomal biogenesis factor 11A) The peptide sequence was selected from the middle region of PEX11A (NP_003838). Peptide sequence MKRVTCDRAKKEKSASQDPLWFSVAEEETEWLQSFLLLLFRSLKQHPPLL.
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP15970520 View Documents Novus BiologicalsSupplier Diversity Partner
NBP15970520UL
20ul Each for $149.00
Add to cart
 
NBP159705 View Documents Novus BiologicalsSupplier Diversity Partner
NBP159705
100 ul Each for $329.00
Add to cart
 
Description

Description

PEX11A Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

PEX11A
Rabbit
Polyclonal
Unconjugated
8800
Affinity Purified
Western Blot
RUO
Synthetic peptides corresponding to PEX11A(peroxisomal biogenesis factor 11A) The peptide sequence was selected from the middle region of PEX11A (NP_003838). Peptide sequence MKRVTCDRAKKEKSASQDPLWFSVAEEETEWLQSFLLLLFRSLKQHPPLL.
Primary
IgG
28 kDa peroxisomal integral membrane protein, hsPEX11p, MGC119947, MGC138534, Peroxin-11A, peroxisomal biogenesis factor 11 alpha, Peroxisomal biogenesis factor 11Aperoxin-11A, peroxisomal membrane protein 11A, PEX11, PEX11-alpha, PMP28, Protein PEX11 homolog alpha
PBS and 2% Sucrose with 0.09% Sodium Azide
Immunogen affinity purified
Store at -20C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit