Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PEX16 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA526158
Description
PEX16 Polyclonal Antibody for Western Blot, IHC (P)
PEX16 is required for peroxisome membrane biogenesis. It may play a role in early stages of peroxisome assembly and can recruit other peroxisomal proteins, such as PEX3 and PMP34, to de novo peroxisomes derived from the endoplasmic reticulum (ER). It may function as receptor for PEX3.
Specifications
PEX16 | |
Polyclonal | |
Unconjugated | |
PEX16 | |
PBD8A; PBD8B; peroxin 16; peroxin-16; Peroxisomal biogenesis factor 16; peroxisomal membrane protein PEX16; peroxisome biogenesis factor 16; Pex16 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
9409 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.44 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q9Y5Y5 | |
PEX16 | |
KLH conjugated synthetic peptide between 247-275 amino acids from the central region of human PEX16. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction