Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PEX19 Mouse anti-Human, Mouse, DyLight 755, Clone: 554, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243544IR

Catalog No. NBP243544IR


Add to cart

Description

Description

PEX19 Monoclonal antibody specifically detects PEX19 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

PEX19
554
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
PEX19
Recombinant protein encompassing a sequence within the center region of human PEX19. The exact sequence is proprietary.
Protein G purified
Store at 4C in the dark.
Monoclonal
Human, Mouse
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
D1S2223Ehousekeeping gene, 33kD, HK3333 kDa housekeeping protein, Peroxin-19, peroxisomal biogenesis factor 19, Peroxisomal farnesylated proteinFLJ55296, PMP1, PMPI, PXFperoxin-19, PXMP1
Mouse
IgG2a
0.1 ml
Primary
5824
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit