Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PEX6 Mouse anti-Human, Rat, HRP, Clone: S233-8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242199H

Catalog No. NBP242199H


Add to cart

Description

Description

PEX6 Monoclonal antibody specifically detects PEX6 in Human, Rat samples. It is validated for Western Blot.
Specifications

Specifications

PEX6
S233-8
Western Blot
Purified
PEX6
Fusion protein amino acids 2-294 (N-terminus) of human PEX6. Rat: 82% identity (243/293 amino acids identical). Mouse: 82% identity (244/294 amino acids identical). <50% identity with other PEX proteins.
Protein G purified
Store at 4C in the dark.
Monoclonal
Less then 50% identity with other PEX proteins.Detects approx 100kDa.
Western Blot
HRP
PBS with No Preservative
PAF2, PAF-2peroxisome assembly factor 2, Peroxin-6, peroxisomal biogenesis factor 6peroxin-6, Peroxisomal-type ATPase 1, peroxisome biogenesis factor 6, PXAAA1peroxisomal AAA-type ATPase 1
Mouse
IgG1
0.1 ml
Primary
5190
Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit