Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PEX6 Mouse anti-Human, Mouse, Rat, Clone: S233-8, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527670

Catalog No. PIMA527670


Add to cart

Description

Description

PEX6 encodes a member of the AAA (ATPases associated with diverse cellular activities) family of ATPases. This member is a predomitly cytoplasmic protein, which plays a direct role in peroxisomal protein import and is required for PTS1 (peroxisomal targeting signal 1, a C-terminal tripeptide of the sequence ser-lys-leu) receptor activity. Mutations in this gene cause peroxisome biogenesis disorders of complementation group 4 and complementation group 6.
Specifications

Specifications

PEX6
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.1% sodium azide; pH 7.4
P54777, Q13608, Q99LC9
PAF 2, Peroxin 6, Peroxisomal AAA type ATPase 1, Peroxisomal biogenesis factor 6, PXAAA1, peroxisomal-type ATPase 1, peroxisome biogenesis factor 6
Mouse
IgG1
100 μg
-20°C
Primary
117265, 224824, 5190
Immunocytochemistry, Immunofluorescence, Western Blot
S233-8
Unconjugated
PEX6
Liquid
Pex6
Fusion protein amino acids 2-294 (N-terminus) of human PEX6. Rat: 82% identity (243/293 amino acids identical). Mouse: 82% identity (244/294 amino acids identical). <50% identity with other PEX proteins.
Protein G
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit