Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PEX7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561908
Description
Immunogen sequence: MSAVCGGAAR MLRTPGRHGY AAEFSPYLPG RLACATAQHY GIAGCGTLLI LDPDEAGLRL FRSFDWNDGL FDVTWSENNE HVLITCSGD Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 80%.
This gene encodes the cytosolic receptor for the set of peroxisomal matrix enzymes targeted to the organelle by the peroxisome targeting signal 2 (PTS2). Defects in this gene cause peroxisome biogenesis disorders (PBDs), which are characterized by multiple defects in peroxisome function. There are at least 14 complementation groups for PBDs, with more than one phenotype being observed in cases falling into particular complementation groups. Although the clinical features of PBD patients vary, cells from all PBD patients exhibit a defect in the import of one or more classes of peroxisomal matrix proteins into the organelle. Defects in this gene have been associated with PBD complementation group 11 (PBD-CG11) disorders, rhizomelic chondrodysplasia punctata type 1 (RCDP1), and Refsum disease (RD).
Specifications
PEX7 | |
Polyclonal | |
Unconjugated | |
PEX7 | |
MmPEX7; PBD9B; PCDP1; Peroxin 7; Peroxin-7; peroxisomal biogenesis factor 7; peroxisomal PTS2 receptor; peroxisomal targeting signal 2 receptor; peroxisome biogenesis factor 7; peroxisome targeting signal 2 receptor; Pex7; PEX7 protein; PTS2 receptor; PTS2R; RCDP1; RD | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5191 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O00628 | |
PEX7 | |
Recombinant protein corresponding to Human PEX7. Recombinant protein control fragment (Product #RP-94691). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction