Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PF 04691502, Tocris Bioscience
Potent and selective dual PI 3-K/mTOR inhibitor
Manufacturer: Tocris Bioscience 4820/25
This item is not returnable.
View return policy
Description
Potent and selective dual ATP-competitive PI 3-K/mTOR inhibitor (Ki values are 1.6, 1.8, 1.9, 2.1 and 16 nM for human PI 3-K δ, α, γ, β, and mTOR, respectively). Displays no significant inhibitory activity at more than 80 protein kinases (concentration ≥ 10 μM) including hVps34, PI 3-K downstream kinases, and MAPK family members. Orally available. Induces robust cell cycle arrest at the G1 phase in U87MG cancer cells and antitumor activity in SKOV3 ovarian cancer xenograft models.Specifications
2-Amino-8-[trans-4-(2-hydroxyethoxy)cyclohexyl]-6-(6-methoxy-3-pyridinyl)-4-methyl-pyrido[2,3-d]pyrimidin-7(8H)-one | |
>98% | |
Observed MW: 425.48 |
1013101-36-4 | |
C22H27N5O4 | |
25mg |
Safety and Handling
Recommended Storage : Store at -20°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title