Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PF 04885614, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent NaV1.8 inhibitor (IC50 = 53 nM for human NaV1.8 channel). Exhibits selectivity for hNaV1.8 over hNaV1.6, hNaV1.7, hNaV1.1, hNaV1.2 and hNaV1.5 (IC50 values are 4.2, 7.0, 11, 16 and 27 μM respectively). Orally bioavailable.
Specifications
Quantity | 50 mg |
Formula Weight | 285.26 |
Percent Purity | >98% |
Chemical Name or Material | PF 04885614 |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction