Learn More
Tocris Bioscience™ PF 05175157 Available on GSA/VA Contract for Federal Government customers only.
Acetyl-CoA carboxylase (ACC) 1 and 2 inhibitor
$166.40 - $686.50
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
57-901-0 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
5790/10
Each for $166.40
Add to cart
57-905-0 View Documents Tocris Bioscience™
5790/50
Each for $686.50
Add to cart
Description
Description
Acetyl-CoA carboxylase (ACC) 2 and 1 inhibitor (IC50 values are 45 and 98 nM, respectively). Reduces hepatic and skeletal muscle malonyl-CoA levels in rats. Also inhibits hepatic de novo lipogenesis and reduces whole body respiratory exchange ratio in rats. Orally bioavailable.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.