Learn More
Medchemexpress LLC PF-06263276 | 1421502-62-6 | 98.3% | 566.63 g/mol | C31H31FN8O2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10102450MG
Encompass_Preferred
PF-06263276 is a small-molecule pan-Janus kinase (pan-JAK) inhibitor supplied for preclinical and in vitro research. It reduces pSTAT3 and TYK2 pathway activation and is provided with analytical documentation for quality verification. Not for human or veterinary use.
  • Inhibits JAK1, JAK2, JAK3, and TYK2 with reported low-nanomolar activity.
  • Documented molecular weight 566.63 g/mol and chemical formula C31H31FN8O2.
  • CAS number 1421502-62-6 for unambiguous identification.
  • High purity reported (~98.3% by data sheet) with supporting analytical data.
  • Available in small research quantities, including a 50 mg unit.
  • Accompanied by certificate of analysis and safety data sheet for handling and compliance.
  • Intended for in vitro and preclinical research use only.
Catalog No. 50-002-12037