Learn More
Tocris Bioscience™ PF 3084014 hydrobromide
Potent γ-secretase inhibitor
$173.90 - $734.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
57-511-0 View Documents Tocris Bioscience™
5751/10
Each for $173.90
Add to cart
57-515-0 View Documents Tocris Bioscience™
5751/50
Each for $734.00
Add to cart
Description
Description
Potent γ-secretase inhibitor (IC50 values are 1.2 and 6.2 nM in whole cell and cell-free assays, respectively). Reduces Aβ in brain, CSF and plasma in mice and guinea pigs.