Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PF 3716556

$199.00 - $829.00

Specifications

Assay Percent Range >98%
Formula Weight Observed MW: 394.47
Chemical Name or Material 8-[[(4R)-3,4-Dihydro-5-methyl-2H-1-benzopyran-4-yl]amino]-N-(2-hydroxyethyl)-N,2-dimethyl-imidazo[1,2-a]pyridine-6-carboxamide
Molecular Formula C22H26N4O3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
41-911-0 View Documents Tocris Bioscience™
4191/10
10mg Each for $199.00
Add to cart
 
41-915-0R View Documents Tocris Bioscience™
4191/50
50mg Each for $829.00
Add to cart
 
Description

Description

H+,K+-ATPase inhibitor (pIC50 = 6 in human recombinant ion-leaky assays); more potent in acidic conditions. Highly selective for H+,K+-ATPase in vitro; displays no activity at Na+,K+-ATPase. Also displays selectivity for H+,K+-ATPase over a range of 50 receptors and ion channels (IC50 > 10 μM). Inhibits gastric acid secretion in rat and dog models.
Specifications

Specifications

>98%
Observed MW: 394.47
8-[[(4R)-3,4-Dihydro-5-methyl-2H-1-benzopyran-4-yl]amino]-N-(2-hydroxyethyl)-N,2-dimethyl-imidazo[1,2-a]pyridine-6-carboxamide
C22H26N4O3
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit