Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PF 429242, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Reversible, competitive inhibitor of sterol regulatory element-binding protein (SREBP) site 1 protease (IC50 = 0.175 μM). Selective for site 1 protease against a panel of serine proteases. Inhibits rate of cholesterol synthesis in CHO cells (IC50 = 0.53 μM). Also displays antiviral activity. Cell permeable.
Specifications
Quantity | 10 mg |
Formula Weight | 482.49 |
Percent Purity | >97% |
Chemical Name or Material | PF 429242 |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction