Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PF 4778574, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
This item is not returnable. View return policy
1219633-99-4
C19H22N2O3S2
390.516
FFAGHPLLBXWCSF-MSOLQXFVSA-N
unii-ti55d82z4b,2-propanesulfonamide, n-3r,4s-3-4-5-cyano-2-thienyl phenyl tetrahydro-2h-pyran-4-yl,n-3r,4s-3-4-5-cyano-2-thienyl phenyl tetrahydro-2h-pyran-4-yl-2-propanesulfonamide
44462786
N-[(3R,4S)-3-[4-(5-cyanothiophen-2-yl)phenyl]oxan-4-yl]propane-2-sulfonamide
CC(C)S(=O)(=O)NC1CCOCC1C2=CC=C(C=C2)C3=CC=C(S3)C#N
This item is not returnable. View return policy
1219633-99-4
C19H22N2O3S2
390.516
FFAGHPLLBXWCSF-MSOLQXFVSA-N
unii-ti55d82z4b,2-propanesulfonamide, n-3r,4s-3-4-5-cyano-2-thienyl phenyl tetrahydro-2h-pyran-4-yl,n-3r,4s-3-4-5-cyano-2-thienyl phenyl tetrahydro-2h-pyran-4-yl-2-propanesulfonamide
44462786
N-[(3R,4S)-3-[4-(5-cyanothiophen-2-yl)phenyl]oxan-4-yl]propane-2-sulfonamide
CC(C)S(=O)(=O)NC1CCOCC1C2=CC=C(C=C2)C3=CC=C(S3)C#N

Positive allosteric modulator of AMPA receptors

Positive allosteric modulator of AMPA receptors (Ki = 85 nM). Prevents ketamine-induced working memory impairments. Brain penetrant.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 390.52
Percent Purity >98%
Chemical Name or Material PF 4778574

Recommended Storage : Store at +4°C