Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris™ PF 514273

Potent and selective CB1 antagonist

Manufacturer:  Tocris™ 3676/50

 View more versions of this product

Catalog No. 36-765-0R


  Check Availability
Add to cart

Disclaimers:

For Research Use Only


Specifications

Description

Potent and selective CB1 receptor antagonist (Ki values are 1 and > 10000nM at CB1 and CB2 receptors respectively). Inhibits food intake in vivo following oral administration.
  • Chemical Name: 2-(2-Chlorophenyl)-3-(4-chlorophenyl)-7-(2,2-difluoropropyl)-6,7-dihydro-2H-pyrazolo[3,4-f][1,4]oxazepin-8(5H)-one
  • Formula: C21H17Cl2F2N3O2
  • CAS Number: 851728-60-4
  • Solubility: Soluble to 100mM in DMSO and to 50mM in ethanol
  • Purity: >98
Description & Specifications

Specifications

PF 514273
>98
50mg
PF 514273
452.28
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit