Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PF 6274484

High affinity and potent covalent EGFR kinase inhibitor

$195.00 - $819.00

Specifications

Chemical Name or Material N-[4-[(3-Chloro-4-fluorophenyl)amino]-7-methoxy-6-quinazolinyl]-2-propenamide
Molecular Formula C18H14ClFN4O2
CAS 1035638-91-5
Product Type Target: EGF Receptor Inhibitors
Formula Weight 372.78
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
59-395-0 View Documents Tocris Bioscience™
5939/50
50mg Each for $819.00
Add to cart
 
59-391-0 View Documents Tocris Bioscience™
5939/10
10mg Each for $195.00
Add to cart
 
Description

Description

High affinity and potent covalent EGFR kinase inhibitor (Ki = 0.14 nM). Inhibits autophosphorylation of WT and mutant EGFR in tumor cells (IC50 values are 5.8 and 6.6 nM, respectively).
Specifications

Specifications

N-[4-[(3-Chloro-4-fluorophenyl)amino]-7-methoxy-6-quinazolinyl]-2-propenamide
1035638-91-5
372.78
C18H14ClFN4O2
Target: EGF Receptor Inhibitors
Safety and Handling

Safety and Handling

Recommended Storage : Store at −20°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit