Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PF 670462, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent and selective casein kinase 1ε (CK1ε) and CK1δ inhibitor (IC50 values are 7.7 and 14 nM respectively) that displays > 30-fold selectivity over 42 other common kinases. Inhibits PER protein nuclear translocation (EC50 = 290 nM) causing phase shifts in circadian rhythms and attenuates methamphetamine-stimulated locomotion in vivo.
Specifications
Quantity | 10 mg |
Formula Weight | 410.32 |
Percent Purity | >99% |
Chemical Name or Material | PF 670462 |
Safety and Handling
Recommended Storage | Desiccate at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction