Learn More
PF 750, Tocris
Selective FAAH inhibitor
Manufacturer:  R&D Systems 3307/50
Quantity 50mg
 View more versions of this product
Catalog No. 33-075-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>98%
345.44
PF 750
C22H23N3O
N-Phenyl-4-(3-quinolinylmethyl)-1-piperidinecarboxamide
Specifications
50mg
>98%
345.44
PF 750
959151-50-9
C22H23N3O
N-Phenyl-4-(3-quinolinylmethyl)-1-piperidinecarboxamide
Safety and Handling
Recommended Storage : Store at Room Temperature