PF 8380, Tocris™

Manufacturer: R&D Systems 4078/50

Quantity 50mg

 View more versions of this product

Catalog No. 40-785-0

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1144035-53-9
    Molecular Formula C22H21Cl2N3O5
    Formula Weight 478.33
    Synonym 3, 5-Dichlorobenzyl-[4-[3-oxo-3-(2-oxo-2, 3-dihydrobenzoxazol-6-yl)propyl]]piperazine-1-carboxylate
    Chemical Name or Material PF 8380

    Specifications

    Formula Weight 478.33
    Quantity 50mg
    Molecular Formula C22H21Cl2N3O5
    CAS 1144035-53-9
    Synonym 3, 5-Dichlorobenzyl-[4-[3-oxo-3-(2-oxo-2, 3-dihydrobenzoxazol-6-yl)propyl]]piperazine-1-carboxylate
    Recommended Storage Store at -20°C
    Chemical Name or Material PF 8380

    Store at -20°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit