Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PF 9184

Potent microsomal prostaglandin E synthase 1 (mPGES-1) inhibitor

$199.00 - $839.00

Specifications

Chemical Name or Material N-[3′,4′-Dichloro(1,1′-biphenyl)-yl]-4-hydroxy-2H-1,2-benzothiazine-3-carboxamide 1,1-dioxide
Molecular Formula C21H14Cl2N2O4S
CAS 1221971-47-6
Product Type Target: Additional Isomerase Inhibitors
Formula Weight 461.32
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
59-171-0 View Documents Tocris Bioscience™
5917/10
10mg Each for $199.00
Add to cart
 
59-175-0 View Documents Tocris Bioscience™
5917/50
50mg Each for $839.00
Add to cart
 
Description

Description

Potent microsomal prostaglandin E synthase 1 (mPGES-1) inhibitor (IC50 values are 16.5 and 1080 nM for human and rat mPGES-1, respectively). Exhibits >6500-fold selectivity for mPGES-1 over COX-1 and COX-2. Inhibits IL-1β-induced PGE2 synthesis in vitro.
Specifications

Specifications

N-[3′,4′-Dichloro(1,1′-biphenyl)-yl]-4-hydroxy-2H-1,2-benzothiazine-3-carboxamide 1,1-dioxide
1221971-47-6
461.32
C21H14Cl2N2O4S
Target: Additional Isomerase Inhibitors
Safety and Handling

Safety and Handling

Recommended Storage : Store at −20°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit