Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PF4 Monoclonal Antibody (170138)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA523926

Catalog No. PIMA523926


Only null left
Add to Cart

Description

Description

In Western blots, no cross-reactivity with recombinant mouse (rm) CXCL6, recombinant human (rh) CXCL6, rhCXCL7, rmCXCL13 or rhCCL21 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

Platelet factor-4 is a 70-amino acid protein that is released from the alpha-granules of activated platelets and binds with high affinity to heparin. Its major physiologic role appears to be neutralization of heparin-like molecules on the endothelial surface of blood vessels, thereby inhibiting local antithrombin III activity and promoting coagulation. As a strong chemoattractant for neutrophils and fibroblasts, PF4 probably has a role in inflammation and wound repair (Eisman et al., 1990).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PF4
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
P02776
Pf4
E. coli-derived recombinant human CXCL4/PF4 Glu32-Ser101.
100 μg
Primary
Human
Antibody
IgG2b
Flow Cytometry, Western Blot
170138
Unconjugated
Pf4
chemokine (C-X-C motif) ligand 4; C-X-C motif chemokine 4; Cxcl4; Endothelial cell growth inhibitor; Iroplact; oncostatin-A; PF4; PF-4; Pf4a; Platelet factor 4; Platelet factor 4, short form; RATPF4A; Scyb4
Mouse
Protein A/G
RUO
5196
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.