Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PFAS Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5107211
Description
Sequence homology: Cow: 83%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 92%; Rabbit: 79%
PFAS Polyclonal antibody specifically detects PFAS in Human samples. It is validated for Western Blot.Specifications
PFAS | |
Polyclonal | |
Unconjugated | |
PFAS | |
4432409B16Rik; FGAM synthase; FGAM synthetase; FGAMS; FGAR amidotransferase; FGARAT; FGAR-AT; Formylglycinamide ribonucleotide amidotransferase; formylglycinamide ribotide amidotransferase; formylglycinamide ribotide synthetase; Gm18; Kiaa0361; PFAS; Phosphoribosylformylglycinamidine synthase; phosphoribosylformylglycinamidine synthase (FGAR amidotransferase); pur4; PURL; Sofa | |
Rabbit | |
Affinity Chromatography | |
RUO | |
5198 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
O15067 | |
PFAS | |
Synthetic peptide located within the following region: ESIMSTQESS NPNNVLKFCD NSSAIQGKEV RFLRPEDPTR PSRFQQQQGL | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction