Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PFDN2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549224

Catalog No. PA549224

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with bovine and rat based on sequence homology.

Binds specifically to cytosolic chaperonin (c-CPN) and transfers target proteins to it. Binds to nascent polypeptide chain and promotes folding in an environment in which there are many competing pathways for nonnative proteins.
Specifications

Specifications

PFDN2
Polyclonal
PBS with 0.09% sodium azide
Q9UHV9, O70591
Prefoldin subunit 2, PFDN2, PFD2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
18637, 5202
Western Blot
Unconjugated
PFDN2
Liquid
PFDN2
KLH conjugated synthetic peptide between 97-125 amino acids from the C-terminal region of human PFDN2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit