Learn More
PFDN2 Rabbit anti-Human, DyLight 488, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP300279G
Catalog No. NB065300
Add to cart
Description
Description
PFDN2 Polyclonal antibody specifically detects PFDN2 in Human samples. It is validated for Western Blot, ELISA
Specifications
Specifications
PFDN2
DyLight 488
50mM Sodium Borate
PFD2, prefoldin 2, prefoldin subunit 2
Produced in rabbits immunized with purified, recombinant PFDN2 (Q9UHV9; Met1-Ser154).
Affinity purified
RUO
Primary
5202
ELISA, Western Blot
Western Blot, ELISA
Purified
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Polyclonal
Human