Learn More
PFDN2 Rabbit anti-Human, DyLight 594, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP300279DL594
Catalog No. NB065298
Add to cart
Description
Description
PFDN2 Polyclonal specifically detects PFDN2 in Human samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
PFDN2
DyLight 594
50mM Sodium Borate
PFD2, prefoldin 2, prefoldin subunit 2
Produced in rabbits immunized with purified, recombinant PFDN2 (Q9UHV9; Met1-Ser154).
Affinity purified
RUO
Primary
5202
Western Blot, ELISA
Western Blot, ELISA
Purified
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Polyclonal
Human