Learn More
PFDN5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 150781AP
Catalog No. 50-172-9958
Add to cart
Description
Description
Binds specifically to cytosolic chaperonin (c-CPN) and transfers target proteins to it. Binds to nascent polypeptide chain and promotes folding in an environment in which there are many competing pathways for nonnative proteins. Represses the transcriptional activity of MYC.
Specifications
Specifications
PFDN5
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q99471, Q9WU28
C Myc binding protein Mm 1, MM 1, MM1, Myc modulator 1, PFD5, PFDN5, prefoldin subunit 5
Rabbit
IgG
150 μL
-20°C
Primary
300257, 5204, 56612
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
PFDN5
Liquid
PFDN5
PFDN5 Fusion Protein Ag7150
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat