Learn More
Selleck Chemical LLCSupplier Diversity Partner PFI-1 (PF-6405761) 50mg 1403764-72-6  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S121650MG
PFI-1 (PF-6405761) is a highly selective BET (bromodomain-containing protein) inhibitor for BRD4 with IC50 of 0.22 μM and for BRD2 with IC50 of 98 nM in a cell-free assay.
Catalog No. 50-710-7