Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PFKFB1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549832
Description
The antibody detects endogenous levels of total PFKFB1 protein.
PFKFB1 Polyclonal antibody specifically detects PFKFB1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
PFKFB1 | |
Polyclonal | |
Unconjugated | |
PFKFB1 | |
6-biphosphatase 1; 6PF-2-K/Fru-2,6-P2ase 1; 6PF-2-K/Fru-2,6-P2ASE liver isozyme; 6-phosphofructo-2-kinase; 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 1; 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase 1; F6PK; Fructose-2,6-bisphosphatase; fructose-6-phosphate,2-kinase:fructose-2,6-bisphosphatase; HL2K; PFK/FBPase 1; PFK-2/FBPase-2 gene A; Pfkfb01; PFKFB1; PFRX | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18639, 24638, 5207 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P07953, P16118, P70266 | |
PFKFB1 | |
Synthesized peptide derived from internal of human PFKFB1/4. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction