Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PG-9 maleate, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

155649-00-6
C21H26BrNO6
468.344
VBUSCULTLLOSLM-SGCJHOTMSA-N
Tropanyl 2-(4-bromophenyl)propionate maleate
56972155

Presynaptic cholinergic modulator

A potent analgesic and nootropic agent. Increases release of acetylcholine.
SMILES CC(C1=CC=C(C=C1)Br)C(=O)OC2CC3CCC(C2)N3C.C(=CC(=O)O)C(=O)O
IUPAC Name (Z)-but-2-enedioic acid;[(1R,5S)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] 2-(4-bromophenyl)propanoate
Formula Weight 468.34
Quantity 10mg
Chemical Name or Material PG-9 maleate
Grade >98%

Recommended Storage : Store at Room Temperature