Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGBD1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550283

Catalog No. PA550283


Add to cart

Description

Description

The antibody detects endogenous levels of total PGBD1 protein.

The piggyBac family of proteins, found in diverse animals, are transposases related to the transposase of the canonical piggyBac transposon from the moth, Trichoplusia ni. This family also includes genes in several genomes, including human, that appear to have been derived from the piggyBac transposons. This gene belongs to the subfamily of piggyBac transposable element derived (PGBD) genes. The PGBD proteins appear to be novel, with no obvious relationship to other transposases, or other known protein families. This gene product is specifically expressed in the brain, however, its exact function is not known.
Specifications

Specifications

PGBD1
Polyclonal
Unconjugated
PGBD1
Liquid
PGBD1
Fusion protein corresponding to residues near the C terminal of human piggyBac transposable element derived 1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
0.8 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Q96JS3
SCAND4, HUCEP-4, dJ874C20.4
Rabbit
IgG
100 μL
-20°C
Primary
84547
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit