Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PGCP Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP158037
Description
PGCP Polyclonal specifically detects PGCP in Human samples. It is validated for Western Blot.Specifications
PGCP | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
Q9Y646 | |
CPQ | |
Synthetic peptides corresponding to PGCP(plasma glutamate carboxypeptidase) The peptide sequence was selected from the N terminal of PGCP. Peptide sequence VDTVGPRLSGSKNLEKAIQIMYQNLQQDGLEKVHLEPVRIPHWERGEESA. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Bovine: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Chicken: 78%. | |
Human, Mouse, Rat, Porcine, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
aminopeptidase, EC 3.4.17, EC 3.4.17.-, plasma glutamate carboxypeptidase | |
Rabbit | |
Affinity Purified | |
RUO | |
10404 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title