Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ PGD Human Recombinant Protein
Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity
$241.00 - $566.50
Specifications
Concentration | 1mg/mL |
---|---|
For Use With (Application) | SDS-PAGE |
Formulation | 20mM Tris-HCl buffer (pH8.0) containing 1mM DTT, 0.1M Nacl, 10% glycerol |
Gene ID (Entrez) | 5226 |
Molecular Weight (g/mol) | TMW: 55.3kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2522382
|
Novus Biologicals™
NBP2522380.02MG |
0.02mg |
Each for $241.00
|
|
|||||
NBP25223801
|
Novus Biologicals™
NBP2522380.1MG |
0.1mg |
Each for $566.50
|
|
|||||
Description
A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-483 of Human PGD The Recombinant Human PGD Protein is derived from E. coli. The Recombinant Human PGD Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.Specifications
1mg/mL | |
20mM Tris-HCl buffer (pH8.0) containing 1mM DTT, 0.1M Nacl, 10% glycerol | |
TMW: 55.3kDa | |
6PGD, EC 1.1.1.44,6-phosphogluconate dehydrogenase, decarboxylating, PGDH, phosphogluconate dehydrogenase | |
Specific activity is > 10 units/mg, which one unit oxidize 1.0 umole of 6-phospho-D-gluconate to D-ribulose 5-phosphate per minute at pH 8.0 at 25C, the presence of beta-NADP. | |
Recombinant |
SDS-PAGE | |
5226 | |
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. | |
PGD | |
Unconjugated | |
>90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain |
For research use only and is not approved for use in humans or in clinical diagnosis
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title