Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGD Mouse anti-Human, Clone: 2A5, Abnova™

Mouse monoclonal antibody raised against full length recombinant human PGD.

Manufacturer:  Abnova Corporation MAB13003

Catalog No. 89-934-762


Add to cart

Description

Description

6-phosphogluconate dehydrogenase is the second dehydrogenase in the pentose phosphate shunt. Deficiency of this enzyme is generally asymptomatic, and the inheritance of this disorder is autosomal dominant. Hemolysis results from combined deficiency of 6-phosphogluconate dehydrogenase and 6-phosphogluconolactonase suggesting a synergism of the two enzymopathies. [provided by RefSeq]

Specifications

Specifications

PGD
2A5
Mouse monoclonal antibody raised against full length recombinant human PGD.
In PBS, pH 7.3 (50% glycerol, 1% BSA, 0.02% sodium azide).
6PGD
Mouse
IgG1
50 uL
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunofluorescence (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot (1:2000) The optimal working dilution should be determined by the end user.
Liquid
PGD
Recombinant protein corresponding to full length human PGD.
Affinity purification
RUO
Primary
5226
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit