Learn More
Enzo Life SciencesSupplier Diversity Partner PGE2 CLIA kit (1x96wells)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADI910001
The PGE2 CLIA kit is a sensitive chemiluminescent competitive enzyme immunoassay kit for the quantitative determination of PGE2 in biological fluids from any species. Alternative Name: Prostaglandin E2. Sensitivity: 6.03 pg/ml (range 7.81 - 1,000 pg/ml). Assay Time: 3 hours. Applications: Chemiluminescence. Species Reactivity: Species independent. Contents: White GxM IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, CLIA Substrate
Catalog No. 50-658-638