Learn More
GE Healthcare pGEX 3' Sequencing Primer, 5'-d[CCG-GGAGCTGCATGTGTCAGAGG]-3 Available on GSA/VA Contract for Federal Government customers only.
Ready for immediate use. GE Healthcare pGEX 3' Sequencing Primer, 5'-d[CCG-GGAGCTGCATGTGTCAGAGG]-3 is a vector primer for sequencing double-stranded DNA inserted into pGEX vectors using the Glutathione S-transferase (GST) Gene Fusion System.
Manufacturer:  GE Healthcare 27141101
Catalog No. 45-002-405
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
The Glutathione S-transferase (GST) Gene Fusion System is a versatile system for the expression, purification and detection of GST-tagged proteins produced in E. coli. The system consists of three major components: pGEX plasmid expression vectors, products for GST purification, and a variety of GST detection products. The pGEX Sequencing Primer is a vector primer for use with the GST system. It is ready for immediate use in sequencing double-stranded DNA inserted into pGEX vectors.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
The Glutathione S-transferase (GST) Gene Fusion System from GE Healthcare is a versatile system for the expression, purification, and detection of GST-tagged proteins produced in E. coli.
pGEX 3' Sequencing Primer, 5'-d[CCG-GGAGCTGCATGTGTCAGAGG]-3
0.05 A 260 unit