Learn More
PGI™ FireLine™ Wildland Coat
Maximum ease of movement with minimum bulk.
Manufacturer:  PGISupplier Diversity Partner 5501872LL
 View more versions of this product
Catalog No. 19-120-3090
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Zipper
UL, NFPA 1977
Large
4.0
Long
255.14g
4 in. High alpine style
Yellow
Indura™ Ultra soft™ Fabric
42 to 44 in.
9 oz.