Learn More
PGI™ FireLine™ Wildland Jumpsuits
Maximum ease of movement with minimum bulk.
Manufacturer:  PGISupplier Diversity Partner 6500272LS
 View more versions of this product
Catalog No. 19-120-3126
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
NFPA 1977
Includes: Radio pocket, microphone holder, cell phone pocket, D-ring loop, CAL OSHA label
42 to 44 in.
Alpine
Large
13.7 to 14.4cm
35 to 38 in.
Short
Zipper
Nomex™ IIIA
Collared
Yellow
Designed to allow maximum ease of movement with minimum bulk
5.4 to 5.7 in.
88.9 to 96.5cm
Back elastic