Learn More
PGI™ FireLine™ Wildland Jumpsuits
Maximum ease of movement with minimum bulk.
Manufacturer:  PGISupplier Diversity Partner 6500272LT
 View more versions of this product
Catalog No. 19-120-3138
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
NFPA 1977
Zipper
Radio pocket, microphone holder, cell phone pocket, D-ring loop, CAL OSHA label
Large
Designed to allow maximum ease of movement with minimum bulk
6.1 to 6.4 in.
88.9 to 96.5cm
Back elastic
Alpine
Nomex™ IIIA
Yellow
42 to 44 in.
15.4 to 16.2cm
35 to 38 in.
Tall