Learn More
PGI™ FireLine™ Wildland Jumpsuits
Maximum ease of movement with minimum bulk.
Manufacturer:  PGISupplier Diversity Partner 65018722XLT
 View more versions of this product
Catalog No. 19-120-3122
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
NFPA 1977
Zipper
Radio pocket, microphone holder, cell phone pocket, D-ring loop, CAL OSHA label
2X-Large
Designed to allow maximum ease of movement with minimum bulk
6.1 to 6.4 in.
109.2 to 116.8cm
Back elastic
Alpine
Indura™ Ultra soft™ Fabric
Yellow
50 to 52 in.
15.4 to 16.2cm
43 to 46 in.
Tall